Sodium poly(naphthaleneformaldehyde)sulfonate

Supplier: SPECTRUM CHEMICAL MFG CORP

S1161-25GM S1161-100GM
700015-614EA 111.23 CAD
700015-614 700016-352
Sodium poly(naphthaleneformaldehyde)sulfonate
Sodium poly(naphthaleneformaldehyde)sulfonate
Formula: C₁₀H₈O₃S·CH₂O)n·xNa
Storage Temperature: Ambient
CAS Number: 9084-06-4

Order Now

Specification Test Results

Appearance Beige Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR