Sodium phosphate dibasic, anhydrous ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: di-Sodium hydrogen phosphate anhydrousSodium phosphate dibasicsec-Sodium phosphateDisodium phosphatedi-Sodium hydrogen orthophosphateSodium hydrogen phosphate

S1402-45KGBL S1402-2.5KG S1402-500GM S1402-12KG
75812-290EA 2541.38 CAD
75812-290 75812-294 75812-292 75812-296
Sodium phosphate dibasic, anhydrous ≥98.0% FCC
di-Sodium hydrogen phosphate

Sodium Phosphate, Dibasic, Anhydrous, FCC is an additive used as an antioxidant, sequestrant and a pH regulator with very high buffering capacity widely used in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₂HPO₄
MW: 141.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4

Order Now

Specification Test Results

Assay (Na₂HPO₄); Dried Basis) 98.0 % min.
Arsenic max.3 mg/kg
Fluoride max. 0.005 %
Insoluble Substances max. 0.2 %
Lead max. 4 mg/kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR