Sodium dihydrogen phosphate monohydrate ≥98.0%, granulated FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1397-2.5KG S1397-500GM S1397-12KG S1397-50KGBL
95035-884EA 1135.42 CAD
95035-884 95035-888 75812-280 75812-282
Sodium dihydrogen phosphate monohydrate ≥98.0%, granulated FCC
Sodium dihydrogen phosphate monohydrate

Sodium Phosphate Monobasic, Monohydrate, Granular, FCC is an additive used as an antioxidant, sequestrant and a pH regulator with very high buffering capacity widely used in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: NaH₂PO₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 103.0 %
Loss On Drying 10.0 - 15.0 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.005 %
Insoluble Substances max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR