Sodium dihydrogen phosphate monohydrate, granulated, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

S1396-12KG S1396-2.5KG S1396-2.5KG-CS S1396-500GM S1396-500GM-CS S1396-50KGBL
75812-268EA 4705.32 CAD
75812-268 75812-270 75812-272 75812-274 75812-276 75812-278
Sodium dihydrogen phosphate monohydrate, granulated, BiotechGrade
Sodium dihydrogen phosphate monohydrate

Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.

Formula: NaH₂PO₄·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Granules
Odor Odorless
Assay (NaH₂PO₄) 98.0-103.0 %
Water 10.0- 15.0 %
pH of a 5% Solution 4.1 - 4.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR