Sodium dihydrogen phosphate, anhydrous ≥98.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

SO188-12KG SO188-45KGBL
75812-464EA 2425.85 CAD
75812-464 75812-466
Sodium dihydrogen phosphate, anhydrous ≥98.0% FCC
Sodium dihydrogen phosphate

Sodium Phosphate Monobasic, Anhydrous, FCC is used as a thickening agent and emulsifier in animal feed, toothpaste, and evaporated milk. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: NaH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 103.0 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.005 %
Insoluble Substances max. 0.2 %
Lead (Pb) max. 4 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR