Disodium hydrogen phosphate heptahydrate, granulated, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

S1412-12KG S1412-2.5KG S1412-500GM
75812-298EA 2840.37 CAD
75812-298 75812-300 75812-302
Disodium hydrogen phosphate heptahydrate, granulated, BiotechGrade
di-Sodium hydrogen phosphate heptahydrate

Sodium Phosphate Dibasic, Heptahydrate, Granular, BiotechGrade is used along with disodium hydrogen phosphate in food treatment to adjust pH. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6

Order Now

Specification Test Results

Appearance To Pass Test
Odor Odorless
Assay (Na₂HPO₄) 98.0-100.5 %
Loss On Drying @ 105 C 43.0- 50.0 %
Solubility To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR