Sodium metabisulfite ≥90.0%, granulated FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium pyrosulfite, Disodium disulphite, Sodium disulfite, di-Sodium disulphite, di-Sodium disulfite, Sodium pyrosulphite, Disodiumdisulphite, Disodium disulfite, Disodiumdisulfite, Sodium disulphite

SO181-12KG SO181-50KGBL
75812-454EA 1791.72 CAD
75812-454 75812-456
Sodium metabisulfite ≥90.0%, granulated FCC
Sodium metabisulphite

Sodium Metabisulfite, Granular, FCC is used as a preservative and antioxidant in food and is also known as E223. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formula: Na₂S₂O₅
MW: 190.11 g/mol
Melting Pt: 150
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Na₂S₂O₅) 90.0 - 100.5 %
Iron (Fe) max. 10 mg/kg
Lead max. 2 mg/kg
Selenium (Se) max. 5 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR