Sodium dodecyl sulfate (SDS) FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1332-500GM S1332-12KG S1332-45KGBL
CA95035-846EA 688 CAD
CA95035-846 75812-248 75812-250
Sodium dodecyl sulfate (SDS) FCC
Sodium dodecyl sulphate (SDS)

Sodium Lauryl Sulfate, FCC is a surfactant and emulsifier commonly used in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Skull and crossbones;Exclamation mark
UN2811

Formula: CH₃(CH₂)₁₁OSO₃Na
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 59.0 % min.
Alkalinity To Pass Test(0.2
Sodium Chloride and Sodium Sulfate max. 8.0 %
Lead (Pb) max. 2 mg/kg
Unsulfated Alcohols max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR