Sodium hydroxide, pellets, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

S1301-12KG S1301-2.5KG S1301-2.5KG-CS S1301-500GM S1301-500GM-CS S1301-50KGBL
75812-222EA 1600.54 CAD
75812-222 75812-224 75812-226 75812-228 75812-230 75812-232
Sodium hydroxide, pellets, BiotechGrade
Sodium hydroxide

Sodium Hydroxide, Pellets, BiotechGrade or caustic soda, is an inorganic, highly basic compound that is water soluble, as well as soluble in ethanol and methanol. One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates


GHS hazard : Corrosion;Exclamation mark
UN1823

Formula: NaOH
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystallin
Odor Odorless
Assay (Total Alkali Including A Max. of 3.0% NaCO₃) 95.0 - 100.5 %
Appearance of a 10% Solution Clear and Color
Solubility To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR