tri-Sodium citrate dihydrate ≥99.0%, granulated FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1256-12KG S1256-45KGBL
75812-210EA 2003.12 CAD
75812-210 75812-212
tri-Sodium citrate dihydrate ≥99.0%, granulated FCC
tri-Sodium citrate dihydrate

Sodium Citrate, Dihydrate, Granular, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Na₃C₆H₅O₇·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5 %
Water 10.0 - 13.0 %
Alkalinity To Pass Test
Lead (Pb) max. 2 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR