Trisodium citrate, anhydrous ≥99.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate

S1986-12KG S1986-45KGBL S1986-12KGPL
CA700006-212EA 4808 CAD
CA700006-212 CA700006-214 75812-406
Trisodium citrate, anhydrous ≥99.0% USP
tri-Sodium citrate

Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₆H₅Na₃O₇
MW: 258.07 g/mol
Melting Pt: 150
Flash Pt: 290
Storage Temperature: Ambient
MDL Number: MFCD00012462
CAS Number: 68-04-2
Merck Index: 12,08746

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5 %
Alkalinity To Pass Test
Water Determination max. 1.0 %
Tartrate To Pass Test
Heavy Metals max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR