Trisodium citrate, anhydrous ≥99.0%, powder FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate
Sodium Citrate, Anhydrous, Powder, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₅Na₃O₇ MW: 258.07 g/mol Melting Pt: 150 Flash Pt: 290 Storage Temperature: Ambient |
MDL Number:
MFCD00012462 CAS Number: 68-04-2 Merck Index: 12,08746 |
Specification Test Results
Assay (C₆H₅Na₃O₇; Anhydrous Basis) | 99.0 - 100.5 % |
Lead | max. 2 mg/kg |
Alkalinity | To Pass Test |
Water Determination | max. 1 % |
Identification | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...