Sodium chloride ≥99.0% Ph. Eur., USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Saline, NaCl, Sea salt, Halite
Sodium Chloride, USP, EP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
NaCl MW: 58.44 g/mol Boiling Pt: 1413 Melting Pt: 801 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 Merck Index: 14,08599 |
Specification Test Results
Assay (NaCl; Dried Basis) | 99.0 - 100.5 % |
Appearance of Solution | To Pass Test |
Acidity or Alkalinity | To Pass Test |
Loss On Drying | max. 0.5 % |
Bromides | max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...