Sodium chloride, granulated, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

S1249-12KG S1249-2.5KG-CS S1249-45KGBL S1249-500GM S1249-500GM-CS
75812-194EA 976.44 CAD
75812-194 75812-196 75812-198 75812-200 75812-202 75812-204
Sodium chloride, granulated, BiotechGrade
Sodium chloride

Sodium Chloride, Granular, BiotechGrade is an ionic compound that is also known as salt. It is responsible for the salinity level of the ocean. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance White, Crystalli
Odor Odorless
Assay (Dried Basis) 99.0 - 101.0 %
Loss On Drying @ 105 C max. 0.5 %
Iodide or Bromide To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR