Sodium carbonate, anhydrous ≥99.5% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

S1228-100GM S1228-12KG S1228-250GM S1228-45KGBL
75812-186EA 116.92 CAD
75812-186 75812-188 75812-190 75812-192
Sodium carbonate, anhydrous ≥99.5% FCC
Sodium carbonate

Sodium Carbonate, Anhydrous, FCC is a food additive (E500) used as an acidity regulator, anti-caking agent, raising agent, and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: Na₂CO₃
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.5 - 100.5 %
Lead (Pb) max. 10 mg/kg
Loss On Drying max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR