Sodium bitartrate monohydrate ≥99.5%, BiotechGrade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (2R,3R)-2,3-Dihydroxysuccinic acid, sodium salt hydrate

S1177-500GM S1177-500GM-CS
75812-174EA 931.24 CAD
75812-174 75812-176
Sodium bitartrate monohydrate ≥99.5%, BiotechGrade
Sodium bitartrate monohydrate

Sodium Bitartrate, Monohydrate, BiotechGrade, also known as monosodium tartrate, is a sodium acid salt of tartaric acid and used as an acidity regulator as well as a test for ammonium cation. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: C₄H₅NaO₆·H₂O
MW: 190.083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150038
CAS Number: 6131-98-2

Order Now

Specification Test Results

Assay 99.5 % min.
pH of a 0.1 M Solution @ 20°C 2.5 - 4.0
Specific Rotation +22 +/- 1
Absorbance of a 0.1 M Solution @ 260 NM max. 0.008
Absorbance of a 0.1 M Solution @ 280 NM max. 0.004


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR