Sodium hydrogen carbonate ≥99.7%, powder, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

S1147-12KG S1147-2.5KG S1147-500GM S1147-500GM-CS
75812-162EA 1365.52 CAD
75812-162 75812-164 75812-166 75812-168
Sodium hydrogen carbonate ≥99.7%, powder, BiotechGrade
Sodium hydrogen carbonate

Sodium Bicarbonate, Powder, BiotechGrade is a commonly used laboratory chemical that can react with both acids and bases, therefore it can be used as a neutralizer. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: NaHCO₃
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (NaHCO₃; Dried Basis) 99.7 - 100.3 %
Solubility 1 G in 11 ML H₂O, 25°C Soluble
Ammonia (NH₃) To Pass Test
Calcium max. 0.01 %
Chloride (Cl) max. 0.015 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR