Sodium L(+)-ascorbate ≥99.0%, powder USP

Supplier: Spectrum Chemical Mfg. Corp.

S1349-5KG S1349-500GM S1349-50KGBL
700009-376EA 3145.61 CAD
700009-376 95035-694 75812-256
Sodium L(+)-ascorbate ≥99.0%, powder USP
Sodium L(+)-ascorbate

Sodium Ascorbate, Powder, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₆H₇NaO₆
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0%
pH of a 10% Solution 7.0 - 8.0
Specific Rotation [A]D +103° to +108°
Loss On Drying max. 0.25%
Heavy Metals (As Pb) max. 0.002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR