Sodium L(+)-ascorbate ≥99.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1137-50KGBL S1137-5KG
75812-154EA 5506.06 CAD
75812-154 75812-156
Sodium L(+)-ascorbate ≥99.0% FCC
Sodium L(+)-ascorbate

Sodium Ascorbate, FCC is used as a food additive as an antioxidant and acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₆H₇NaO₆
Storage Temperature: Ambient
MDL Number: MFCD00082340
CAS Number: 134-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d142b281353c28f2a72502b60fece03e926023d&vid=988032859afe97984b0fc1b8bfd43cff093f099a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.0 %
Optical Rotation +103 to +108
Loss On Drying max. 0.25 %
Lead (Pb) max. 2 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR