Sodium acetate trihydrate ≥99.0%, granulated FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acetic acid sodium salt trihydrate

S1114-12KG S1114-2.5KG S1114-500GM S1114-50KGBL
75812-136EA 2064.98 CAD
75812-136 75812-138 75812-140 75812-142
Sodium acetate trihydrate ≥99.0%, granulated FCC
Sodium acetate trihydrate

Sodium Acetate, Trihydrate, Granular, FCC is used as food flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 99.0-101.0 %
Loss On Drying 36.0- 41.0 %
Alkalinity (As Na₂CO₃) max. 0.05 %
Lead (Pb) max. 2 mg/kg
Potassium Compounds To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR