Sodium acetate, anhydrous ≥99.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

S1112-12KG S1112-2.5KG S1112-45KGBL
75812-128EA 2406.94 CAD
75812-128 75812-130 75812-132 75812-134
Sodium acetate, anhydrous ≥99.0% FCC
Sodium acetate

Sodium Acetate, Anhydrous, FCC is used for controlling the pH level of food items and acts as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: H₃CCOONa
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3

Order Now

Specification Test Results

Assay (Dried Basis) 99.0- 101.0 %
Alkalinity (As NaOH) max. 0.2 %
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 1.0 %
Potassium Compounds To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR