Silicon dioxide ≥94.0% FCC 325 Mesh

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Silicon(IV) oxideSilicium (IV) oxideSilicium dioxide

S1388-20KGBL S1388-500GM S1388-2.5KG S1388-12KG
75812-266EA 1990.28 CAD
95035-642 75812-266 95035-644 700000-688
Silicon dioxide ≥94.0% FCC 325 Mesh
Silicium dioxide

Silicon Dioxide, 325 Mesh, FCC.


All Spectrum Chemical FCC grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Specification Test Results

Assay (SiO₂, Ignited) 94.0 % min.
Lead (Pb) max.5 mg/kg
Loss On Drying max. 7.0 %
Loss On Ignition max. 8.5 %
Soluble Ionizable Salts (As Sodium Sulfate) max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR