Saccharin sodium dihydrate ≥99.0%, granulated USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide dihydrate, Sodium Saccharin, Dihydrate, Saccharin sodium salt, o-sulphobenzimide sodium salt dihydrate, Saccharin Sodium Salt Dihydrate, o-Benzoic sulphimide sodium salt, o-sulfobenzimide sodium salt dihydrate, o-Benzoic sulfimide sodium salt

SO195-100GM SO195-12KG
75812-472EA 358.34 CAD
75812-472 75812-474
Saccharin sodium dihydrate ≥99.0%, granulated USP
Saccharin sodium dihydrate

Saccharin Sodium, Dihydrate, Crystalline, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₇H₅NNaO₃S·2H₂O
MW: 242.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151213
CAS Number: 6155-57-3

Order Now

Specification Test Results

Assay (C₇H₄Nnao₃S) 99.0 - 101.0 %
Clarity of Solution To Pass Test
Color of Solution To Pass Test
Acidity or Alkalinity To Pass Test
Water max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR