Riboflavine (vitamin B2) ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

R1032-1KGPL R1032-500GMG R1032-100GMG R1032-25GMGL R1032-25KGBL
95035-550EA 1800.65 CAD
95035-550 95035-554 95035-548 95035-552 75812-114
Riboflavine (vitamin B2) ≥98.0% FCC
Riboflavine (Vitamin B2)

Riboflavin, FCC is also referred to as vitamin B2 and used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₇H₂₀N₄O₆
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation +56.5 to +59.5
Loss On Drying max. 1.5 %
Lumiflavin To Pass Test
Residue On Ignition max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR