Propyl-4-hydroxybenzoate sodium salt ≥94.0% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Propyl parahydroxybenzoate sodium salt, Sodium Propyl Parahydroxybenzoate, Propylparaben Sodium, Propylparaben sodium salt, 4-Hydroxybenzoic acid propyl ester sodium salt, Sodium 4-(propoxycarbonyl)phenolate, Sodium 4-propoxycarbonylphenoxide

P1457-100GM P1457-25GM P1457-50KGBL
75811-962EA 221.52 CAD
75811-962 75811-964 75811-966
Propyl-4-hydroxybenzoate sodium salt ≥94.0% NF
Propyl 4-hydroxybenzoate sodium salt

PROPYLPARABEN SODIUM, NF, also known as propyl paraben, is a natural substance originating in several plants and insects. It is a preservative mostly in cosmetics, lotions, or creams that are water-based and some foods, and pharmaceuticals. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₀H₁₁NaO₃
MW: 202.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016477
CAS Number: 35285-69-9

Order Now

Specification Test Results

Assay (Anhydrous Basis) 94.0 - 102.0 %
Completeness of Solution To Pass Test
pH of a 1 in 1000 Solution 9.5 - 10.5
Water max. 5.0 %
Chloride max. 0.035 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR