Propyl-4-hydroxybenzoate ≥99.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

P1452-12KG P1452-50KGBL
75811-954EA 2979.72 CAD
75811-954 75811-956
Propyl-4-hydroxybenzoate ≥99.0% FCC
Propyl 4-hydroxybenzoate

Propylparaben, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00002354
CAS Number: 94-13-3

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 100.5 %
Acidity To Pass Test
Melting Range 95 - 98 °C
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR