(±)-1,2-Propanediol ≥99.5% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

PR130-20LTPL PR130-4LTPL PR130-4LTPL-CS
CA700002-916EA 12376.99 CAD
CA700002-916 75812-102 75812-104
(±)-1,2-Propanediol ≥99.5% USP
Propylene glycol

Propylene Glycol, USP is used in pharmaceutical products as an excipient solvent in oral and topical drug products. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5 % min.
Specific Gravity @ 25°C 1.035 - 1.037
Acidity To Pass Test
Water Determination max. 0.2 %
Residue On Ignition max. 0.007 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR