(±)-1,2-Propanediol ≥99.5% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Propylene Glycol, FCC is used as a humectant, solvent, and a preservative in food products. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
CH₃CH(OH)CH₂OH Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 |
|
Specification Test Results
Assay | 99.5 % min. |
Acidity | To Pass Test |
Distillation Range | 185 - 189 °C |
Specific Gravity @ 25°C | 1.035- 1.037 |
Lead (Pb) | max. 1 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...