Sulfur ≥99.5%, precipitated

Supplier: Spectrum Chemical Mfg. Corp.

S1384-12KG S1384-2.5KG S1384-500GM S1384-50KGBL
75812-258EA 1975.63 CAD
75812-258 75812-260 75812-262 75812-264
Sulfur ≥99.5%, precipitated
Sulphur

Precipitated Sulfur is an abundant, non-metal, multivalent chemical element. In cosmetics and personal care products Sulfur is used to enhance the appearance and feel of hair, by increasing hair body, suppleness, or sheen, or by improving the texture of hair that has been damaged physically or by chemical treatment and to enhance the appearance of dry or damaged skin by reducing flaking and restoring suppleness


GHS hazard : Flame
UN1350

Formula: S
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
UN: 1350
ADR: 4.1,III

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.5 - 100.5 %
Reaction to Litmus Neutral
Water max. 0.5 %
Residue On Ignition max. 0.3 %
Other Forms of Sulfur To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR