Potassium sorbate ≥98.0%, powder FCC

Supplier: Spectrum Chemical Mfg. Corp.

P1408-12KG P1408-45KGBL
75811-944EA 2691.29 CAD
75811-944 75811-946
Potassium sorbate ≥98.0%, powder FCC
Potassium sorbate

Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CH₃CH=CHCH=CHCOOK
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 101.0 %
Acidity To Pass Test
Alkalinity To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR