di-Potassium hydrogen phosphate, anhydrous ≥98.0% FCC
Supplier: Spectrum Chemical Mfg. Corp.
Potassium Phosphate, Dibasic, Anhydrous, FCC is used as an acidity regulator, antioxidant, sequestrant and stabilizer in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
K₂HPO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 |
|
Specification Test Results
Assay (K₂HPO₄) | 98.0 % min. |
Arsenic (As) | max.3 mg/kg |
Fluoride | max. 10 mg/kg |
Insoluble Substances | max. 0.2 % |
Lead (Pb) | max. 2 mg/kg |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...