Potassium dihydrogen phosphate ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

P1382-12KG P1382-2.5KG P1382-45KGBL P1382-500GM
75811-916EA 2675.84 CAD
75811-916 75811-918 75811-920 75811-922
Potassium dihydrogen phosphate ≥98.0% FCC
Potassium dihydrogen phosphate

Potassium Phosphate Monobasic, FCC is used to stabilize, thicken and regulate the acidity and moisture in foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
Merck Index: 13,07744

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 % min.
Arsenic (As) max.3 mg/kg
Fluoride max. 10 mg/kg
Insoluble Substances max. 0.2 %
Lead (Pb) max. 2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR