di-Potassium hydrogen phosphate, anhydrous, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

P1386-2.5KG P1386-500GM
75811-934EA 2083.63 CAD
75811-934 75811-936
di-Potassium hydrogen phosphate, anhydrous, BiotechGrade
di-Potassium hydrogen phosphate

Potassium Phosphate Dibasic, Anhydrous, BiotechGrade, also known as Potassium monohydrogen phosphate, is used to make buffer solutions. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Formula: K₂HPO₄
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Odor Odorless
Assay (K₂HPO₄) 98.0-100.5 %
pH of a 5% Solution @ 25°C Approx. 9
Solubility Clr.Colorless So
Loss On Drying @ 105 C max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR