Potassium nitrate ≥99.5%, BiotechGrade

75811-914CS 2074.13 CAD
75811-914
Potassium nitrate ≥99.5%, BiotechGrade
Potassium nitrate

Potassium Nitrate, BiotechGrade - this compound has been tested to alleviate NaCl stress symptoms in plants. Mechanistic studies suggest that this may be achieved through reduction of electrolyte leakage, malondialdehyde and soluble sugar contents and improving the activities of antioxidant enzymes. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: KNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.5 % min.
pH of a 1M Solution @ 20°C 5.0-8.0
Absorbance Cut-Off of a 1M Solution 328 NM
Loss On Drying @ 150°C max. 0.1 %
Insoluble Matter max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR