Potassium hydroxide, pellets FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potash caustic, Caustic potash

P1316-12KG P1316-50KGBL
75811-886EA 1742.03 CAD
75811-886 75811-888
Potassium hydroxide, pellets FCC
Potassium hydroxide

Potassium Hydroxide, Pellets, FCC is used as a rinse or chemical peel for fruits and vegetables, a stabilizer to prolong shelf-life, and as a thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Skull and crossbones
UN1813

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Assay (Total Alkali As KOH) 85.0 - 100.5 %
Carbonate (As K₂CO₃) max. 3.5 %
Insoluble Substances To Pass Test
Lead (Pb) max. 2 mg/kg
Mercury max. 0.1 MG/

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR