Potassium chloride ≥99.0%, granular FCC

Supplier: Spectrum Chemical Mfg. Corp.

P1255-12KG P1255-250LBBL P1255-45KGBL
75811-864EA 1275.46 CAD
75811-864 75811-866 75811-868
Potassium chloride ≥99.0%, granular FCC
Potassium chloride

Potassium Chloride, Granular, FCC is used as a flavor enhancer, flavoring agent, nutrient supplement, pH control agent, and stabilizer or thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: KCl
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 % min.
Acidity or Alkalinity To Pass Test
Loss On Drying max. 1.0 %
Heavy Metals (As Pb) max. 5 mg/kg
Iodide or Bromide To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR