Potassium chloride ≥99.0%, granular, BiotechGrade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassiumchloride
Potassium Chloride, Granular, BiotechGrade is commonly used as a laboratory reagent as a standard. It may be used as a calibration standard ionic solution for measuring electrical conductivity because potassium chloride solutions, when prepared carefully, produce repeatable measurable properties. Potassium chloride is also a good source of ionic chloride, which should precipitate insoluble chloride salts upon addition to a solution of an appropriate metal ion.
Formula:
KCl MW: 74.55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 |
Specification Test Results
Assay (KCl) | 99.0-100.5 % |
Acidity / Alkalinity | To Pass Test |
Appearance of a 10% Solution | Clear & Colorles |
Solubility (10 G in 50 ML) | To Pass Test |
Loss On Drying @ 105 C | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...