Potassium hydrogen phthalate ≥99.5%, BiotechGrade

Supplier: Spectrum Chemical Mfg. Corp.

P1192-500GM P1192-500GM-CS
75811-822EA 582.64 CAD
75811-822 75811-824
Potassium hydrogen phthalate ≥99.5%, BiotechGrade
Potassium hydrogen phthalate

Potassium Biphthalate, BiotechGrade, also known as potassium hydrogen phthalate or KHP, is an acidic salt compound used as a primary standard for acid-base titrations and as a buffering agent. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.


Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Formula: HOOCC₆H₄COOK
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (C₈H₅O₄K; Dried Basis) 99.5 % min.
pH of a 0.1M Solution @ 20°C 3.5 - 5.0
Insoluble Matter max. 0.01 %
Chloride max. 0.002 %
Sulfates max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR