Potassium hydrogen carbonate ≥99.0%, granular FCC

Supplier: Spectrum Chemical Mfg. Corp.

P1182-13 P1182-12KG P1182-45KGBL
CA11024-384EA 1297.01 CAD
CA11024-384 75811-814 75811-816
Potassium hydrogen carbonate ≥99.0%, granular FCC
Potassium hydrogen carbonate

Potassium Bicarbonate, Granular, FCC is used as a carbon dioxide source for leavening in baking. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: KHCO₃
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6

Order Now

Specification Test Results

Assay (Dried Basis) 99.0 - 101.5 %
Carbonate (CO₃) To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 0.25 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR