Polyethylene glycol 400 NF

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: PEG 400

PO110-20LT PO110-25ML
CA700011-206EA 4641.99 CAD
CA700011-206 75812-040
Polyethylene glycol 400 NF
Polyethylene glycol 400

Polyethylene Glycol 400, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
Density: 1.128
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

Average Molecular Weight 380 - 420
Completeness and Color of Solution To Pass Test
Viscosity @98.9 C (210 F) +/-0.3 C 6.8 - 8.0 Cs
pH of a 5% Solution @ 25°C 4.5 - 7.5
Residue On Ignition max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR