Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Supplier: Spectrum Chemical Mfg. Corp.
P1813-2.5LTG
P1813-20LT
P1813-500ML
700012-012EA
595.98
CAD
700012-012
700012-014
700012-016
Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Poloxamer 182 (Pluronic® L-62, Synperonic® L62)
Poloxamer 182 belongs to a group of compounds known as poloxamers that are made up of three polymer blocks. In cosmetic and personal care product formulations, it functions as a surfactant - cleansing agent. It can be used in soaps and cleansers.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH Storage Temperature: Ambient |
MDL Number:
MFCD00082049 CAS Number: 9003-11-6 |
Specification Test Results
Cloud Point (10 % Aqueous) | 22 - 26 °C |
pH of a 2.5% Aqueous Solution | 5.0- 7.5 |
Color (APHA) | max. 50 |
Water | max. 0.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...