Orthophosphoric acid NF

Supplier: Spectrum Chemical Mfg. Corp.

PH175-2.5LGL-C PH175-2.5LPV-C PH175-4LTPV-CS PH175-500ML-CS
75812-030CS 1673.09 CAD
75812-030 75812-032 75812-034 75812-036 75812-038
Orthophosphoric acid NF
Orthophosphoric acid

PHOSPHORIC ACID, NF is a also known under the name orthophosphoric acid. A mineral acid that is most often presented as a nonvolatile and clear liquid that has a viscous texture. It has a wide field of uses, such as a buffering agent in aqueous solutions, used as a food additive in many soft drinks and in orthodontics as an etching solution. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - Keep tightly closed


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: H₃PO₄
Density: 1.685
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay (H₃PO₄; By Wt.) 85.0 - 88.0 %
Sulfate (SO₄) To Pass Test
Heavy Metals (As Pb) max. 10 ppm
Alkali Phosphates To Pass Test
Nitrate (NO₃) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR