Orthophosphoric acid 85% ≥85% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Phosphoric acid

P1097-1LT P1097-2.5LT-CS P1097-2.5LTPV P1097-200LTBL P1097-20LTBL P1097-4LT P1097-500ML-CS P1097-500MLGL
75811-780EA 438.2 CAD
75811-780 75811-782 75811-784 75811-786 75811-788 75811-790 75811-792 75811-794 75811-796
Orthophosphoric acid 85% ≥85% FCC
Orthophosphoric acid

Phosphoric Acid, 85 Percent, FCC is used to acidify foods and beverages such as various colas and jams. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Corrosion;Exclamation mark
UN1805

Formula: H₃PO₄
Density: 1.685
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay 85.0 % min.
Arsenic (As) max.3 mg/kg
Cadmium (Cd) max. 3 mg/kg
Fluoride max. 10 mg/kg
Lead (Pb) max. 3 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR