Phenol ≥99.0%, fused crystals USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

PH120-1.5KGPL PH120-25KGBL
75812-026EA 1319.3 CAD
75812-026 75812-028
Phenol ≥99.0%, fused crystals USP
Phenol

PHENOL, FUSED CRYSTAL, USP is extracted from petroleum and mainly used to produce precursors of plastics and is also used as a building block to synthetically produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesic among them. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE; AIR SENSITIVE; MOISTURE SENSITIVE: Keep tightly closed in light-resistant containers. Store under nitrogen


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Density: 1.071
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
UN: 1671
ADR: 6.1 II
Merck Index: 12,07390

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5 %
Clarity of Solution and Reaction To Pass Test
Congealing Temperature 39°C min.
Water Determination max. 0.5 %
Nonvolatile Residue max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR