Petrolatum, white USP

Supplier: Spectrum Chemical Mfg. Corp.

PE125-15.9KG PE125-174KGBL
CA700000-484EA 2072 CAD
CA700000-484 75812-024
Petrolatum, white USP
Petrolatum

Petrolatum, White, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
MDL Number: MFCD00147839
CAS Number: 8009-03-8

Order Now

Specification Test Results

BHT As Reported
Tocopherols As Reported
Specific Gravity @60 C 0.815 - 0.880
Melting Range 38 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR