Peppermint oil FCC, rectified

Supplier: Spectrum Chemical Mfg. Corp.

P1595-100ML P1595-10KGBL P1595-25ML P1595-500MLGL
75811-974EA 308.78 CAD
75811-974 75811-976 75811-978 75811-980
Peppermint oil FCC, rectified
Peppermint oil

Peppermint Oil, Rectified, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Storage Temperature: Ambient
MDL Number: MFCD00147870
CAS Number: 8006-90-4

Order Now

Specification Test Results

Assay (Total Esters As Menthyl Acetate; (C₁₂H₂₂O₂) 5.0 % min.
Assay (Total Menthol; C₁₀H₂₀O) 50.0 % min.
Angular Rotation -18° to -32°
Refractive Index @ 20°C 1.459 - 1.465
Solubility in Alcohol To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR