Olive oil NF

Supplier: SPECTRUM CHEMICAL MFG CORP CA

OL130-20LTPL OL130-500MLG
CA89078-908EA 5755.01 CAD
CA89078-908 CA95034-798
Olive oil NF
Olive oil

OLIVE OIL, NF, is an oil that comes from the olive fruit and is a fat. As an excipient, it is used as a vehicle for oily injection suspensions and as an emollient. Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00131764
CAS Number: 8001-25-0

Order Now

Specification Test Results

Fatty Acid Composition To Pass Test
Sterol Composition To Pass Test
Acid Value max. 0.3
Alkaline Impurities To Pass Test
Absence of Sesame Oil To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR