2-Ethylhexyl-4-methoxycinnamate ≥95.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: 4-Methoxycinnamic Acid 2-Ethylhexyl Ester, 4-Methoxycinnamic Acid Octyl Ester, Octinoxate, Octyl 4-Methoxycinnamate, 2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

O1123-500GM O1123-20KG
CA95034-780EA 1575.01 CAD
CA95034-780 75811-762
2-Ethylhexyl-4-methoxycinnamate ≥95.0% USP
2-Ethylhexyl-4-methoxycinnamate

Octinoxate, USP is a UV-stabilizer excipient. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072582
CAS Number: 5466-77-3

Order Now

Specification Test Results

Assay 95.0 - 105.0 %
Specific Gravity 1.005 - 1.013
Refractive Index 1.542 - 1.548
Chromatographic Purity To Pass Test
Acidity To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR