Nicotinic acid (Niacin) 98.0-102.0% (dried basis), powder USP

Supplier: Spectrum Chemical Mfg. Corp.
Warning

NI100-12KGBL NI100-50KGBL
75811-752EA 1389.06 CAD
75811-752 75811-754
Nicotinic acid (Niacin) 98.0-102.0% (dried basis), powder USP
Nicotinic acid (Niacin)

NIACIN, POWDER, USP, is also referred to as vitamin B3 and nicotine acid and is one of the essential human nutrients. It is a colorless, water soluble derivative of pyridine. As a food supplement it is available over the counter but can also be found naturally in various food groups such as meat, fruit and vegetables and seeds. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.1.0 %
Residue On Ignition max. 0.1 %
Chloride (Cl) max. 0.02 %
Sulfates (SO₄) max. 0.02 %
Cadmium As reported
Lead (Pb) As reported
ARSENIC (As) As reported
Mercury (Hg) As reported
Related compounds To pass test
Identification To pass test
Certified Kosher
Certified Halal
Residual solvents To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR