Sodium dihydrogen phosphate monohydrate 98,0, granulated BP, USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate

SO130-12KG SO130-2.5KG SO130-500GM
89050-380EA 3723.27 CAD
89050-380 95035-890 95035-892 75812-424
Sodium dihydrogen phosphate monohydrate 98,0, granulated BP, USP
Sodium dihydrogen phosphate monohydrate

Monobasic Sodium Phosphate, Monohydrate, Granular, USP, BP is used as a buffering agent and sequestering agent.


All Spectrum Chemical USP, BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (NaH₂PO₄; Anhydrous Basis) 98.0 - 100.5 %
pH of a 5% Solution (Acidity) 4.2 - 4.5
Water Determination 11.5 - 14.5%
Insoluble Substances max. 0.2%
Chloride (Cl) max. 0.014%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR